Skip to Content
Merck
All Photos(2)

Key Documents

G2754

Sigma-Aldrich

Gly-DL-Asp

≥98%, suitable for electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2O5
CAS Number:
Molecular Weight:
190.15
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

Gly-DL-Asp,

Assay

≥98%

form

powder

technique(s)

electrophoresis: suitable

color

white

storage temp.

−20°C

SMILES string

NCC(=O)NC(CC(O)=O)C(O)=O

InChI

1S/C6H10N2O5/c7-2-4(9)8-3(6(12)13)1-5(10)11/h3H,1-2,7H2,(H,8,9)(H,10,11)(H,12,13)

InChI key

SCCPDJAQCXWPTF-UHFFFAOYSA-N

Gene Information

human ... SLC15A1(6564)

Application

Glycyl dipeptides such as glycyl-L-aspartate are useful as model compounds for the development of chiral separation technologies such as chiral ligand-exchange capillary electrophoresis and capillary electrochromatography (CEC).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Chitra et al.
Acta crystallographica. Section C, Crystal structure communications, 63(Pt 1), o11-o13 (2007-01-09)
The title bis(glycyl-L-aspartic acid) oxalate complex {systematic name: bis[2-(2-ammonioacetamido)butanedioic acid] oxalate 0.4-hydrate}, 2C6H11N2O5+.C2O4(2-).4H2O, crystallizes in a triclinic space group with the planar peptide unit in a trans conformation. The asymmetric unit consists of two glycyl-L-aspartic acid molecules with positively charged
E C O'Brien et al.
Journal of inorganic biochemistry, 77(3-4), 135-139 (2000-03-04)
The first solution studies at physiological pH for the formation of metal complexes of taurine, +NH3CH2CH2S03-, one of the most abundant low molecular weight organic compounds in the animal kingdom, are reported. The complexes Cu(Gly-GlyH-1) (1) and [Cu(Gly-AspH-1)] (2) react
R Kaul et al.
Journal of neurochemistry, 56(1), 129-135 (1991-01-01)
Canavan disease, an autosomal recessive disorder, is characterized biochemically by N-acetylaspartic aciduria and aspartoacylase (N-acyl-L-aspartate amidohydrolase; EC 3.5.1.15) deficiency. However, the role of aspartoacylase and N-acetylaspartic acid in brain metabolism is unknown. Aspartoacylase has been purified to apparent homogeneity with
D S Eggleston et al.
International journal of peptide and protein research, 19(2), 206-211 (1982-02-01)
The crystal structure of the acidic dipeptide glycyl-L-aspartic acid dihydrate, Gly-L-Asp X 2H2O, C6H10N2O5 X 2H2O, has been determined by means of three-dimensional counter X-ray data. The dipeptide crystallizes in space group P212121 of the orthorhombic system with four formula
Mohammad K Islam et al.
Biochimica et biophysica acta, 1770(8), 1212-1218 (2007-06-19)
To better understand the mechanism of vanadium reduction in ascidians, we examined the reduction of vanadium(V) to vanadium(IV) by NADPH and the reduction of vanadium(IV) to vanadium(III) by L-cysteine methyl ester (CysME). UV-vis and electron paramagnetic resonance spectroscopic studies indicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service