Skip to Content
Merck
All Photos(1)

Key Documents

F1054

Sigma-Aldrich

Anti-FOXC2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FKHL14, Anti-MFH-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

human, mouse

technique(s)

microarray: suitable
western blot: 1-2 μg/mL using total extracts of NIH3T3-L1 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXC2(2303)
mouse ... Foxc2(14234)

Related Categories

Immunogen

synthetic peptide corresponding to amino acids 439-454 of mouse FOXC2, conjugated to KLH via an N-terminal added lysine residue. The immunizing sequence is conserved in human and mouse. The sequence is partially conserved in FOXC1, another member of Forkhead family.

Application

Anti-FOXC2 antibody produced in rabbit is suitable for microarray and western blotting at a concentration of 1-2μg/mL using total extracts of NIH3T3-L1 cells.

Biochem/physiol Actions

Forkhead box protein C2 (FOXC2) is a protein encoded by the FOXC2 gene in humans. It is a member of the fork head box (FOX) family of transcription factors and acts as a positive transactivator. FOXC2 is expressed in somitic mesoderm, the endocardium and blood vessels as well as the condensing mesenchyme of the bones and kidney and in head mesenchyme. It induces epithelial-to-mesenchymal transition (EMT), which is an important step in Extrahepatic cholangiocarcinoma (EHCC) invasion and metastasis. FOXC2 may act as a promising molecular target for regulating EHCC metastasis. It is mutated in the human vascular disease lymphedema-distichiasis and plays an essential role in lymphatic vascular development. FOXC2 gene expression may provide an effective target for anti-EMT-based therapies for the treatment of claudin-low/basal B breast tumors or other EMT-/CSC-enriched tumors.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Watanabe et al.
Cancer science, 104(11), 1427-1432 (2013-08-08)
Extrahepatic cholangiocarcinoma (EHCC) is a cancer with a poor prognosis, and the postoperative survival of patients depends on the existence of invasion and metastasis. The epithelial-to-mesenchymal transition (EMT) is an important step in EHCC invasion and metastasis. Forkhead box protein
Brett G Hollier et al.
Cancer research, 73(6), 1981-1992 (2013-02-05)
Resistance to chemotherapy and metastases are the major causes of breast cancer-related mortality. Moreover, cancer stem cells (CSC) play critical roles in cancer progression and treatment resistance. Previously, it was found that CSC-like cells can be generated by aberrant activation
K H Kaestner et al.
Development (Cambridge, England), 122(6), 1751-1758 (1996-06-01)
The 'winged helix' or 'forkhead' transcription factor gene family is defined by a common 100 amino acid DNA binding domain which is a variant of the helix-turn-helix motif. Here we describe the structure and expression of the mouse fkh-6 and
N Miura et al.
Genomics, 41(3), 489-492 (1997-05-01)
The very recently found evolutionarily conserved DNA-binding domain of 100 amino acids, termed the fork head domain, emerged from a sequence comparison of the rat hepatocyte transcription factor HNF-3 alpha and the homeotic gene fork head of Drosophila. We previously
Konstantin I Ivanov et al.
Molecular and cellular biology, 33(19), 3749-3761 (2013-07-24)
One of the key mechanisms linking cell signaling and control of gene expression is reversible phosphorylation of transcription factors. FOXC2 is a forkhead transcription factor that is mutated in the human vascular disease lymphedema-distichiasis and plays an essential role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service