Skip to Content
Merck
All Photos(1)

Key Documents

EP5910714009

Adapter for Eppendorf® S-4x400 Rotor

for 175-225 ml bottle, pkg of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

feature

Holds 175 - 225 mL tubes
for 175-225 ml bottle

packaging

pkg of 2 ea

manufacturer/tradename

Eppendorf® 5910714009

technique(s)

centrifugation: suitable

compatibility

for use with 400 mL round bucket

Looking for similar products? Visit Product Comparison Guide

General description

Adapter for various size tubes and bottles for use with Eppendorf S-4x400 rotor.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service