Key Documents
EP5820770007
Adapter for Eppendorf® FA-45-12-17/FA-45-16-17/FA-45-20-17 Rotors
capacity, 1 HPLC tube, pack of 4
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
capacity 1 HPLC tube
packaging
pack of 4
manufacturer/tradename
Eppendorf® 5820770007 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with FA-45-16-17
for use with FA-45-20-17
for use with Rotor FA-45-12-17
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)