Skip to Content
Merck
All Photos(1)

Key Documents

EP5820710004

ROTOR FOR EPPENDORF® 5810/5810R CENTRIFUGES

A-2-DWP-AT, aerosol-tight, for 5810/5810R, with 2 buckets, 2 caps & 2 plate carriers

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

feature

to hold 2 x 1 filtration plate kit
to hold 2 x 6 deepwell plates
to hold 2 x 6 microplates
2 caps & 2 plate carriers
aerosol-tight
deepwell plate rotor
for 5810/5810R
with 2 buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5820710004 (Global)

parameter

3,486 × g max. RCF
4,500 rpm

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

General description

  • Features an aerosol-tight cap that facilitates safe centrifugation of potentially hazardous materials.
  • Rotors are made of anodized aluminum for high chemical resistance and to prevent corrosion.
  • Maximum loading height of 67 mm (89 mm without lid)
  • Both rotor and buckets are autoclavable.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service