EP4861000793
Eppendorf®Xplorer® Plus
24-channel, adjustable, tip volume 5-100 μL, light yellow
Synonym(s):
Eppendorf Xplorer, Xplorer Plus
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
AC/DC input
, universal plug set
feature
adjustable
24-channel
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 4861000793
tip volume
5-100 μL
battery power
lithium
color
light yellow
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
<UL><LI>Multi-functional rocker with up is up and down is down functionality</LI><LI>Selection dial for quick and easy selection of functions</LI><LI>Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position</LI><LI>Spring-loaded tip cone for all pipettes up to 1,000 μL for minimal tip attachment forces helps to reduce stress</LI><LI>Powerful rechargeable battery for up to 8 hours work without the need for recharging</LI><LI>Separate power socket: Work can continue while charging</LI><LI>Adjustment to external conditions (e.g. geographic height, liquids with high density)</LI><LI>Lower part of pipette can be autoclaved to ensure decontamination</LI><LI>9 different languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish</LI><LI>Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals</LI></UL>
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Xplorer is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service