Skip to Content
Merck
All Photos(1)

Key Documents

CLS6520

Corning® multiwell plates, plate lids and sealing mats

96 well polycarbonate PCR plate lid, non-sterile, 25/cs

Synonym(s):

96 multiwell plate lids, 96 well microplate lids, 96 well microtiter plate lids, 96 well plate lids, PCR plate lids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.17
material:
polycarbonate
polycarbonate plate
round wells
size:
96 wells

material

polycarbonate
polycarbonate plate
round wells

sterility

non-sterile

feature

lid

packaging

pack of 1
case of 25

manufacturer/tradename

Corning 6520

technique(s)

PCR: suitable

size

96 wells

color

clear

suitability

suitable for (cell cuture)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Polycarbonate lids are thermally stable up to 135°C and can be used on Thermowell 96 well polycarbonate plates (fits CLS6510, CLS6511 and CLS6514) with oil overlays
  • Lids are intended to be covers only and do not prevent evaporation

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service