Skip to Content
Merck
All Photos(1)

Key Documents

CLS431252

Corning® Erlenmeyer (Fernbach design) cell culture flasks

3 L Erlenmeyer (Fernbach Design) Flask w/ Vent Cap, polycarbonate, sterile, 4/cs

Synonym(s):

Corning Erlenmeyer flasks, cell culture flasks, conical flasks, plastic Erlenmeyer flasks, shaker flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
eCl@ss:
32040501
NACRES:
NB.13
Pricing and availability is not currently available.

material

flat bottom flask
polycarbonate
polypropylene cap (vented, Fernbach design)
vent cap
wide-mouth
wide-mouth neck

description

RNase- and DNase-free

sterility

sterile

feature

cap
disposable
Flask style: Erlenmeyer (Fernbach design)

packaging

pack of 1
case of 4

manufacturer/tradename

Corning 431252

flask capacity

3 L

color

orange cap

suitability

suitable for (Ideal for shaker culture applications)

Looking for similar products? Visit Product Comparison Guide

General description

  • Fernbach design increases surface area for gas exchange
  • Made from optically-clear polycarbonate
  • Ideal for shaker culture applications
  • Vent caps supplied in every case of product for applications requiring sterile gas exchange
  • Sterilized by gamma radiation and certified nonpyrogenic

Other Notes

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A SARS-CoV-2 spike ferritin nanoparticle vaccine protects hamsters against Alpha and Beta virus variant challenge
Kathryn McGuckin Wuertz, et al.
Nature (2019)
Mammalian cell transient expression, non-affinity purification, and characterization of human recombinant IGFBP7, an IGF-1 targeting therapeutic protein
Man-Shiow Jiang, et al.
International Immunopharmacology (2021)
Jinsung Hong et al.
Biotechnology progress, 35(5), e2831-e2831 (2019-05-03)
An alternating tangential flow (ATF) perfusion-based transient gene expression (TGE) bioprocess has been developed using human embryonic kidney (HEK) 293 cells to produce H1-ss-np, a promising candidate for a universal influenza vaccine. Two major adjustments were taken to improve the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service