Skip to Content
Merck
All Photos(2)

Key Documents

CLS3897BC

Corning® 96 Well Microplate With Generic Barcode

V-bottom clear, polystyrene, bag of 25 ×, non-sterile, lid: no

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.24
material:
V-bottom clear
polystyrene
polystyrene
well volume:
320 μL
sterility:
non-sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid: no

material

V-bottom clear
polystyrene
polystyrene

sterility

non-sterile

feature

lid: no

packaging

pack of 10
case of 50
bag of 25 ×

manufacturer/tradename

Corning 3897BC

well volume

320 μL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service