CLS3821
Corning® 384 well microplate, low volume
black polystyrene, flat bottom, non-sterile, lid: no, pack of 10
Synonym(s):
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates
About This Item
black polystyrene
flat bottom
skirt
Recommended Products
material
black bottom
black polystyrene
flat bottom
sterility
non-sterile
feature
lid: no
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3821
size
384 well
well volume
50 μL
well working volume
5-40 μL
wells
384 wells , low volume
suitability
suitable for (molecular, validated for the FLIPR reader)
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Round well design with a solid black flat bottom
- 2.8 mm distance between well bottoms to plate bottom
- Validated by Molecular Devices for the FLIPR reader. Outer wells aligned for proper viewing with FLIPR
- Ideal for miniaturized 384 well fluorescent homogeneous assays
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm2)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service