Skip to Content
Merck
All Photos(1)

Key Documents

JVM-2

96090515, human unknown/unspecified, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human unknown/unspecified
growth mode:
Suspension
karyotype:
46,XX,-8,+t(3;8)(p12-13;q12),t(11;14)(q13;q32)
morphology:
Lymphoblastoid
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

JVM-2, 96090515

biological source

human unknown/unspecified

growth mode

Suspension

karyotype

46,XX,-8,+t(3;8)(p12-13;q12),t(11;14)(q13;q32)

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian B-prolymphocytic leukaemia, EBV-transformed

Cell Line Description

JVM-2 has been derived from EBV and TPA treated mononuclear cells from a 63 year old female patient with B-prolymphocytic leukaemia. Thirty-three percent of cells were shown to be positive for surface lambda light chain, 37% for IgA, 29% for IgG and 3% for IgM and IgD. In addition, 50% were found to express cytoplasmic IgM and lambda light chain. Expression of the following leukocyte differentiation antigens was found: CD9, CD19, CD20, CD23, CD24, MA6, CD37, CD38, FMC1, FMC7, MHC Class I, FMC16, MHC Class II DR/DP/DQ. The immunoglobulin heavy-and light-chain genes showed the same pattern of rearrangement as in the original prolymphocytes. The cells are lymphoblastoid with large floating aggregates present. The cell line has been eradicated from mycoplasma at ECACC.

Application

Ig expression of neoplastic cells

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11,13
D16S539: 12,13
D5S818: 11,12
D7S820: 10,11
THO1: 6,9
TPOX: 8,11
vWA: 17

Culture Medium

RPMI 1640 +2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service