My-La CD8+
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 95051033, human lymph, Lymphoblastoid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human lymph
packaging
tube of 5 μg 95051033-DNA-5UG
pkg of vial of cells 95051033-1VL
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblastoid
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian T Lymphocyte, mycosis fungoides.
Cell Line Description
The cell line My-La, CD8+, also known as My-La, marker chromosome was established from a plaque biopsy of an 82 year old Caucasian patient with mycosis fungoides stage II. Together with My-La, CD8+ a second cell line has been established (My-La, CD4+ Sigma Catalogue number. 95051032) by including IL-2 and IL-4 in the culture medium. My-La, CD8+ shows characteristic chromosome markers, express ß-13 of the T-cell receptor and are double homozygous for HLA-A1.B8. Phenotypically a weak staining was observed for CD2, CD3, CD8, CD25, CD71 and TCR-1 α / ß. There has been no evidence of HTLV-I infection in the cell line and serum from the patient did not demonstrate antibodies against HTLV-I and HTLV-II. My-La, CD8+ cells show a low viability after resuscitation from frozen and a long lag time phase until exponential growth is reached.
Application
Study of antigen-independent proliferation of T- cell lyphoma from mycosis fungoides patient.
Culture Medium
RPMI 1640 + 2mM Glutamine + 100 u/ml IL-2 + 10% human AB seum
Subculture Routine
Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C. Split only 1:2 as maximum dilution: freeze in growth medium plus 10% DMSO, low viability on resuscitation. Use of human serum produces high levels of debris in cultures.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service