COR-L105
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 92031918, human lung, Epithelial-like
Synonym(s):
CORL105 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human lung
packaging
tube of 5 μg 92031918-DNA-5UG
pkg of vial of cells 92031918-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian lung adenocarcinoma
Cell Line Description
Derived from the pleural effusion of a Caucasian male. Cells grow partially attached and in suspension.
Application
COR-L105 cell line has been used to identify chromosomal aberrations by using multicolor fluorescent in situ hybridization (M-FISH) and comparative genomic hybridization (CGH).
DNA Profile
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 9,11
D13S317: 10,11
D16S539: 11,12
D5S818: 11,12
D7S820: 8,11
THO1: 8,9.3
TPOX: 8,11
vWA: 16,17
CSF1PO: 9,11
D13S317: 10,11
D16S539: 11,12
D5S818: 11,12
D7S820: 8,11
THO1: 8,9.3
TPOX: 8,11
vWA: 16,17
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 3-6x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Cells grow in colonies and do not form a confluent monolayer.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service