Fao Cell Line from rat
89042701, rat liver, Hepatocyte
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
Fao Cell Line from rat, rat hepatoma, 89042701
biological source
rat liver
growth mode
Adherent
karyotype
Not specified
morphology
Hepatocyte
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat hepatoma
Cell Line Description
A differentiated cell line derived from H4-11-E-C3 (Sigma catalogue no. 85061112). A versatile tool for studying regulation of mono-oxygenase activities and analysing their role in the activation of xenobiotics e.g. carcinogens, drugs and pesticides.
Application
Differentiation and enzymatic studies
Culture Medium
Coon′s modified Ham′s F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) or kaign′s modified Ham′s F12 + 45mg/l asorbic acid +18mg/1 inositol + 10% FBS +2mM Glutamine
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service