NS0-Turbodoma
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
mouse blood
packaging
tube of 5 μg 06020202-DNA-5UG
pkg of vial of cells 06020202-1VL
growth mode
Not specified
karyotype
Not specified
morphology
lymphoblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse myeloma, serum-free, without cholesterol
Cell Line Description
NS0 cell line (Sigma Catalogue no. 85110503) adapted to grow in the chemically defined protein- and peptide-free medium Turbodoma (Cell Culture Technologies). Cholesterol is not required as a supplement for this culture. This cell line is suitable for use as a fusion parent for serum-free generation of hybridomas. The NS0 cell line is the property of the Medical Research Council (MRC). ECACC is unable to supply this cell line or derivatives of this cell line to commercial entities unless a license has been first secured from LifeArc (formerly known as MRC Technology). Requests for commercial use of this cell line will be forwarded to LifeArc.
Culture Medium
Turbodoma medium (Cell Culture Technologies) + 2mM L-Glutamine (G7513). When freezing cells down use TurboDoma™ medium (50:50 conditioned medium:fresh medium) + 2mM L-Glutamine (G7513) + 10% DMSO.
Subculture Routine
Viability may be poor on resuscitation and may initially decrease further. Full recovery may take up to 2 weeks. A centrifugation step to remove the cryoprotectant is essential. Rapidly thaw the frozen ampoule in a water bath at 37°C for 1-2 minutes. Tran
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Legal Information
TurboDoma is a trademark of Cell Culture Technologies
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service