Skip to Content
Merck
All Photos(5)

Key Documents

AV37678

Sigma-Aldrich

Anti-KCNAB2 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Potassium voltage-gated channel, shaker-related subfamily, β member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

guinea pig, dog, rat, bovine, horse, human, mouse, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNAB2(8514)

General description

Potassium voltage-gated channel subfamily A regulatory beta subunit 2 (KCNAB2), an ion channel gene, codes for a voltage?gated K+ channel β?subunit protein, Kvβ2. This gene is located on human chromosome 1p36. Kvβ2 is expressed ubiquitously in the human heart. It belongs to the aldo?keto reductase superfamily. This member is one of the beta subunits, which are auxiliary proteins associating with functional Kv-α subunits.

Immunogen

Synthetic peptide directed towards the middle region of human KCNAB2

Biochem/physiol Actions

Potassium voltage-gated channel subfamily A regulatory β subunit 2 (KCNAB2) gene is involved in neuroendocrine conditions KCNAB2 serves as a candidate gene for epilepsy. A cardiac arrhythmia disorder, called Brugada syndrome occurs due to the gain-of-function mutation in KCNAB2 gene. Voltage-gated potassium (Kv) channels play a key role in modulating the contraction of smooth muscle, secretion of insulin, the release of neurotransmitters, and excitability of the neuron. It also participates in heart rate, epithelial electrolyte transport, and cell volume.

Sequence

Synthetic peptide located within the following region: WGGKAETERGLSRKHIIEGLKASLERLQLEYVDVVFANRPDPNTPMEETV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles Ashton et al.
Journal of neurosurgery, 1-7 (2020-02-29)
Prior profiling of the human pituitary adenoma (PA) DNA methylome showed the potassium channel subunit-encoding gene KCNAB2 to be highly differentially methylated between nonfunctional PAs (NFPAs) and growth hormone (GH)-secreting PAs, with greater KCNAB2 methylation detected in secretory PAs. KCNAB2
Vincent Portero et al.
Journal of the American Heart Association, 5(6) (2016-06-12)
The Brugada syndrome is an inherited cardiac arrhythmia associated with high risk of sudden death. Although 20% of patients with Brugada syndrome carry mutations in SCN5A, the molecular mechanisms underlying this condition are still largely unknown. We combined whole-exome sequencing
H A Heilstedt et al.
Epilepsia, 42(9), 1103-1111 (2001-10-03)
Clinical features associated with chromosome 1p36 deletion include characteristic craniofacial abnormalities, mental retardation, and epilepsy. The presence and severity of specific phenotypic features are likely to be correlated with loss of a distinct complement of genes in each patient. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service