Skip to Content
Merck
All Photos(1)

Key Documents

AV100810

Sigma-Aldrich

Anti-GTF2F1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-General transcription factor IIF, polypeptide 1, 74 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
guinea pig, dog, human, rabbit
citations:
3
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58 kDa

species reactivity

guinea pig, dog, human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GTF2F1(2962)

Immunogen

Synthetic peptide directed towards the N terminal region of human GTF2F1

Application

Anti-GTF2F1 antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

GTF2F1 is a transcription factor that binds to RNA polymerase II and recruits the initiation complex for transcription. It collaborates with TFIIB and promotes transcription elongation.

Sequence

Synthetic peptide located within the following region: MAALGPSSQNVTEYVVRVPKNTTKKYNIMAFNAADKVNFATWNQARLERD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zengshu Xing et al.
Journal of translational medicine, 19(1), 492-492 (2021-12-05)
Prostate cancer (PCa) belongs to an epithelial malignancy that occurs in the prostate gland and is the most common malignancy of the male genitourinary system. Referring to related literature, circSERPINA3 has been reported to be up-regulated in PCa. However, its
T Aso et al.
Nature, 355(6359), 461-464 (1992-01-30)
At least six chromatographically resolvable general transcription factors may participate in accurate initiation by RNA polymerase II in HeLa cell-derived systems. TFIIF (also termed FC, RAP30/74 and beta/gamma) can bind directly to RNA polymerase II in solution and decrease the
A Finkelstein et al.
Nature, 355(6359), 464-467 (1992-01-30)
RAP30/74 (also known as TFIIF, beta gamma and FC is one of several general factors required for initiation by RNA polymerase II. The small RAP30 subunit of RAP30/74 binds directly to polymerase and appears structurally and functionally homologous to bacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service