Skip to Content
Merck
All Photos(1)

Documents

12731C

Sigma-Aldrich

Porcine Serum

USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

pig sera, pig serum, porcine sera

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

biological source

Porcine

Quality Level

sterility

sterile-filtered

composition

hemoglobin, ≤50 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤50 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Application

Porcine Serum has been used as a negative control to block fluorescein isothiocyanate (FITC) labeled hepatitis C virus (HCV) virus-like particles (VLP) in flow cytometry assay.

Preparation Note

Collected from adult animals.

Analysis Note

Endotoxin tested

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service