Skip to Content
Merck
All Photos(1)

Documents

30709

Sigma-Aldrich

Nitric acid

puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

for determinations with dithizone
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor pressure

8 mmHg ( 20 °C)

Assay

≥65%

form

liquid

ign. residue

≤0.0005% (as SO4)

pH

<1.0

bp

120.5 °C (lit.)

density

1.37-1.41 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 mg/kg
phosphate (PO43-): ≤0.5 mg/kg
sulfate (SO42-): ≤0.5 mg/kg

cation traces

Ag: ≤0.01 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.01 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.1 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.1 mg/kg
Ge: ≤0.05 mg/kg
Hg: ≤0.01 mg/kg
K: ≤0.05 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.01 mg/kg
Sr: ≤0.01 mg/kg
Ti: ≤0.1 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.01 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.1 mg/kg

SMILES string

O[N+]([O-])=O

suitability

passes test for suitability for det. with dithizone

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A yellow coloration of the bottle has no influence whatsoever on the product quality.
Nitric acid (HNO3) is a highly corrosive strong mineral acid. 10%(v/v) nitric acid solution is useful for cleaning laboratory glasswares. Reaction of HNO3 with various original and synthetic mineral dust/mineral oxide surfaces was studied in a low-pressure Knudsen reactor operating at 298K.

Application

Nitric acid (HNO3) may be used for the removal of transition metal catalyst from the single-walled carbon nanotubes (SWNTs) during the purification of SWNTs. It may be used in the trace determinations of Pb and Cd in natural waters by flame atomic absorption spectrometry.

Other Notes

The article number 30709-4X2.5L will be discontinued. Please order the single bottle 30709-2.5L which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneous reactivity of gaseous nitric acid on Al2O3, CaCO3, and atmospheric dust samples: A Knudsen cell study.
Hanisch F and Crowley JN.
The Journal of Physical Chemistry A, 105(13), 3096-3106 (2001)
Nitric acid purification of single-walled carbon nanotubes.
Hu H, et al.
The Journal of Physical Chemistry B, 107(5), 13838-13842 (2003)
Eduardo Carasek et al.
Talanta, 56(1), 185-191 (2008-10-31)
The need for highly reliable methods for the determination of trace elements has been recognised in analytical chemistry and environmental science. A method for the trace analysis of Pb and Cd in natural waters is described. In a preconcentration step
Alyson M Baergen et al.
Environmental science & technology, 47(2), 815-820 (2012-12-15)
The fate of NO(x) (=NO + NO(2)) is important to understand because NO(x) is a significant player in air quality determination through its role in O(3) formation. Here we show that renoxification of the urban atmosphere may occur through the
Stijn Smulders et al.
Toxicological sciences : an official journal of the Society of Toxicology, 141(1), 132-140 (2014-06-14)
The unique physical and chemical properties of nanomaterials have led to their increased use in many industrial applications, including as a paint additive. For example, titanium dioxide (TiO2) engineered nanoparticles (ENPs) have well-established anti-UV, self-cleaning, and air purification effects. Silver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service