Skip to Content
Merck
All Photos(1)

Documents

16037

Sigma-Aldrich

Pyridine

≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

899 °F

expl. lim.

12.4 %

impurities

≤0.01% non-volatile matter
≤0.1% water (Karl Fischer)

refractive index

n20/D 1.509 (lit.)

bp

115 °C (lit.)

mp

−42 °C (lit.)

solubility

water: soluble

density

0.978 g/mL at 25 °C (lit.)

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine is a basic N-heterocyclic compound. It acts as a nitrogen donor ligand and forms many metal-pyridine complexes. Its complexes having tetrahedral and octahedral geometries can be differentiated by infra red spectral investigations.

Application

Pyridine (py) may be used in synthesis of vanadium oxide nanoparticles and iron benzenedicarboxylates, Fe(OH)(BDC)(py)0.85 (BDC = 1,4-benzenedicarboxylate). It may be employed as a catalyst in the Knoevenagel condensation reaction.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porous monodisperse V2O5 microspheres as cathode materials for lithium-ion batteries.
Wang S, et al.
Journal of Materials Chemistry, 21(17), 6365-6369 (2011)
The far-infrared spectra of metal-halide complexes of pyridine and related ligands.
Clark RJH and Williams CS.
Inorganic Chemistry, 4(3), 350-357 (1965)
Metal-organic frameworks based on iron oxide octahedral chains connected by benzenedicarboxylate dianions.
Whitfield TR, et al.
Solid State Sciences, 7(9), 1096-1103 (2005)
An investigation of Knoevenagel condensation reaction in microreactors using a new zeolite catalyst.
Zhang X, et al.
Applied Catalysis A: General, 261(1), 109-118 (2004)
Total syntheses of complanadines A and B.
Le Zhao et al.
Angewandte Chemie (International ed. in English), 52(6), 1722-1725 (2013-01-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service