Skip to Content
Merck
All Photos(1)

Key Documents

SFRVN

SAFC

Sf-RVN Insect Cell Line

Synonym(s):

Sf9 rhabdovirus-negative (Sf-RVN) Insect Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

description

Sf9 rhabdovirus-negative (Sf-RVN) Insect Cell Line

solubility

DMSO: 10%

shipped in

liquid nitrogen

storage temp.

−196°C

General description

Sf-RVN(R)cells are provided to customers in vials containing 1 mL at 10 x 106 cells/mL. Cells are banked in EX-CELL® CD Insect Cell Medium and 10% DMSO.
The Sf-RVN Insect Cell Line is a rhabdovirus-negative Sf9 cell line that can be grown in both adherent and suspension culture. Sf-RVN Insect Cell Line has been adapted for growth in chemically defined medium to be used for the production of recombinant proteins, adeno-associated viruses (AAV) and viral-like particles (VLP). The cell line was tested for adventitious agents and banked under GMP conditions. A technical user guide, with detailed protocols for optimal performances, is provided with the cell line.The EX-CELL® CD Insect Cell Medium, recommended for use, is optimized to get excellent growth and productivity of the Sf-RVN Insect Cell Line. Combined, these two products form the Sf-RVN Platform.

Cell Line Origin

Sf-RVN is an insect cell line originally derived from ovarian tissue of Spodoptera frugiperda (Sf) pupae. The parental line, IPLB-SF-21 (renamed IPLB-SF-21 AE after adaptation into TC-100 medium), was established by J. Vaughn at the Insect Pathology Beltsville in Maryland, USA in the late 1960s. A subclone, designated Sf9, was established in 1983 by G. Smith and C. Cherry. Sf-RVN was isolated from Sf9 by GlycoBac, LLC.

Physical form

Sf-RVN cells are provided to customers in vials containing 1 mL at 10 x 106cells/mL. Cells are banked in EX-CELL® CD Insect Cell Medium and 10% DMSO

Other Notes

Sf-RVN is an insect cell line originally derived from ovarian tissue of Spodoptera frugiperda (Sf) pupae. The parental line, IPLB-SF-21 (renamed IPLB-SF-21 AE after adaptation into TC-100 medium), was established by J. Vaughn at the Insect Pathology Beltsville in Maryland, USA in the late 1960s. A subclone, designated Sf9, was established in 1983 by G. Smith and C. Cherry. Sf-RVN was isolated from Sf9 by GlycoBac, LLC.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
Sf-RVN is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

See how the Sf9 rhabdovirus-free cell line was developed and how we’ve developed a companion chemically-defined insect cell media for protein and viral expression.

See how the Sf9 rhabdovirus-free cell line was developed and how we’ve developed a companion chemically-defined insect cell media for protein and viral expression.

See how the Sf9 rhabdovirus-free cell line was developed and how we’ve developed a companion chemically-defined insect cell media for protein and viral expression.

See how the Sf9 rhabdovirus-free cell line was developed and how we’ve developed a companion chemically-defined insect cell media for protein and viral expression.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service