Skip to Content
Merck
All Photos(1)

Key Documents

PHR1517

Supelco

Dihydrostreptomycin Sulfate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Dihydrostreptomycin sesquisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H41N7O12 · 3/2H2SO4
CAS Number:
Molecular Weight:
730.71
Beilstein:
3894221
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. D1954000
traceable to USP 1203008

API family

dihydrostreptomycin

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@@H](O[C@@H](C)[C@]2(O)CO)O[C@@H]3[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]3NC(N)=N.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)[C@]5(O)CO)O[C@@H]6[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]6NC(N)=N

InChI

1S/2C21H41N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*5-18,26,29-36H,3-4H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,21+;;;/m00.../s1

InChI key

CZWJCQXZZJHHRH-YZTFXSNBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydrostreptomycin sulfate is a semi-synthetic broad-spectrum aminoglycoside antibiotic produced commonly by the reduction of streptomycin. It is used primarily in the treatment of tuberculosis.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Dihydrostreptomycin sulfate may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB9924 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A synthesis of dihydrostreptomycin
Yamasaki T, et al.
The Journal of Antibiotics, 31(12), 1233-1237 (1978)
Veterinary use and antibiotic resistance
Teuber M
Current Opinion in Microbiology, 4(5), 493-499 (2001)
Use of experimental design to optimize a triple-potential waveform to develop a method for the determination of streptomycin and dihydrostreptomycin in pharmaceutical veterinary dosage forms by HPLC-PAD
Martinez MM, et al.
Journal of Pharmaceutical and Biomedical Analysis, 104(5), 81-89 (2015)
Artemis K Sarri et al.
Journal of chromatography. A, 1122(1-2), 275-278 (2006-06-30)
A novel method for the non-derivatization liquid chromatographic determination of streptomycin (STR) and dihydrostreptomycin (DHSTR) was developed and validated based on evaporative light scattering detection (ELSD). Utilizing a ThermoHypersil BetaBasic C18 analytical column, evaporation temperature of 50 degrees C and

Articles

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service