Skip to Content
Merck
All Photos(1)

Key Documents

PHR1407

Supelco

Pamoic Acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Pamoic acid, 1,1′-Methylene-bis(2-hydroxy-3-naphthoic acid), 4,4′-Methylenebis(3-hydroxy-2-naphthoic acid), Embonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16O6
CAS Number:
Molecular Weight:
388.37
Beilstein:
901319
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1493000

API family

oxantel, cycloguanil

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

≥300 °C (dec.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)c1cc2ccccc2c(Cc3c(O)c(cc4ccccc34)C(O)=O)c1O

InChI

1S/C23H16O6/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29/h1-10,24-25H,11H2,(H,26,27)(H,28,29)

InChI key

WLJNZVDCPSBLRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Pamoic Acid may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA2471 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of mebendazole and pyrantel pamoate from tablets by high performance liquid chromatography-reverse phase (RP-HPLC)
Argekar AP, et al.
Talanta, 44(11), 1959-1965 (1997)
Development and validation of a RP-HPLC method for quality control of oxantel pamoate, pyrantel pamoat and praziquantel in tablets
Tatar E, et al.
Marmara Pharmaceutical Journal, 19(1), 27-35 (2015)
Sara Montminy Paquette et al.
Pharmaceutical research, 31(8), 2065-2077 (2014-02-22)
Long-acting injectables (LAIs) are increasingly recognized as an effective therapeutic approach for treating chronic conditions. Many LAIs are formulated to create a poorly soluble depot from which the active agent is delivered over time. This long residing depot can cause

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service