Skip to Content
Merck
All Photos(3)

Key Documents

N5625

Sigma-Aldrich

Naphthol AS phosphate

>99% purity (TLC), powder

Synonym(s):

3-Hydroxy-2-naphthanilide phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14NO5P
CAS Number:
Molecular Weight:
343.27
Beilstein:
2772810
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Naphthol AS phosphate, >99% (TLC), histochemical substrate

Quality Level

Assay

>99% (TLC)

form

powder

solubility

ethanol: 50 mg/mL, clear, light yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

OP(O)(=O)Oc1cc2ccccc2cc1C(=O)Nc3ccccc3

InChI

1S/C17H14NO5P/c19-17(18-14-8-2-1-3-9-14)15-10-12-6-4-5-7-13(12)11-16(15)23-24(20,21)22/h1-11H,(H,18,19)(H2,20,21,22)

InChI key

KVIYXIWBXOQZDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Histochemical substrate for acid and alkaline phosphatase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Primary human alveolar type II epithelial cell chemokine release: effects of cigarette smoke and neutrophil elastase.
Witherden IR
American Journal of Respiratory Cell and Molecular Biology, 30, 500-500 (2004)
Enumerating T-cell subsets on blood smears. An evaluation of an indirect immunoalkaline phosphatase method.
Wong GK
American Journal of Clinical Pathology, 86, 756-756 (1986)
Umut Gazi et al.
The Journal of biological chemistry, 286(10), 7822-7829 (2011-01-06)
The mannose receptor (MR) is an endocytic type I membrane molecule with a broad ligand specificity that is involved in both hemostasis and pathogen recognition. Membrane-anchored MR is cleaved by a metalloproteinase into functional soluble MR (sMR) composed of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service