Skip to Content
Merck
All Photos(1)

Key Documents

BCR168

Picene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1912414
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
pharmaceutical (small molecule)

format

neat

storage temp.

room temp

SMILES string

c1ccc2c(c1)ccc3c2ccc4c5ccccc5ccc34

InChI

1S/C22H14/c1-3-7-17-15(5-1)9-11-21-19(17)13-14-20-18-8-4-2-6-16(18)10-12-22(20)21/h1-14H

InChI key

GBROPGWFBFCKAG-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR168

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K L Platt et al.
Carcinogenesis, 11(10), 1721-1726 (1990-10-01)
The carcinogenic activity of the two polycyclic aromatic hydrocarbons (PAHs), picene (benzo[a]chrysene) and dibenz[a,h]anthracene (DBA), was determined in NMRI mice by five different experimental protocols in order to find out if picene is a carcinogen as predicted by recent quantum
Andrew P Rutter et al.
Environmental science & technology, 43(20), 7770-7777 (2009-11-20)
Carbonaceous atmospheric particulate matter (PM25) collected in the midwestern United States revealed that soot emissions from incomplete coal combustion were important sources of several organic molecular markers used in source apportionment studies. Despite not constituting a major source of organic
Alberto Girlando et al.
Physical chemistry chemical physics : PCCP, 14(5), 1694-1699 (2011-12-24)
The paper reports a complete analysis of the phonon structure of crystalline picene, a recently announced organic semiconductor. Both lattice and intramolecular vibrations are investigated. An exhaustive assignment of lattice phonons is obtained through polarized Raman spectra assisted by lattice
K L Platt et al.
Chemico-biological interactions, 66(3-4), 157-175 (1988-01-01)
Picene, a polycyclic aromatic hydrocarbon (PAH) of environmental relevance has recently been predicted to be carcinogenic, based on quantum mechanical calculation, although in several animal studies no carcinogenicity could be detected. In order to find out if the metabolism of
F J Zhang et al.
The Journal of organic chemistry, 65(13), 3952-3960 (2000-06-24)
A new synthetic approach to polycyclic aromatic compounds is described that entails in the key steps double Suzuki coupling of PAH bisboronic acid derivatives with o-bromoaryl aldehydes to furnish aryl dialdehydes that are converted to larger polycyclic aromatic ring systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service