Skip to Content
Merck
All Photos(2)

Key Documents

90606

Supelco

Methyl heptadecanoate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Heptadecanoic acid methyl ester, Methyl margarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15COOCH3
CAS Number:
Molecular Weight:
284.48
Beilstein:
1782681
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

152-153 °C/0.05 mmHg (lit.)

mp

29.8-30.3 °C (lit.)

application(s)

petroleum

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3

InChI key

HUEBIMLTDXKIPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Methyl heptadecanoate may be used as an internal standard in the gas chromatographic analysis (GC) of an animal fat like lard. It may also be used as internal standard for the gas chromatographic coupled with mass spectrometric (GC-MS) analysis of algal biomass and oils, used in the production of biodiesel.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Optimization of transesterification of animal fat ester using response surface methodology
Jeong T-G, et al.
Bioresource Technology, 100, 25-30 (2009)
Optimization of direct conversion of wet algae to biodiesel under supercritical methanol conditions
Patil.DP, et al.
Bioresource Technology, 102, 118-122 (2011)
Yingyang Zhang et al.
Journal of food science, 79(9), E1695-E1703 (2014-09-13)
The thermal properties of cured meat are important for determining storage life and nutritional quality. However, few studies have focused on the thermal properties of dry-cured pork, particularly in relation to salt level and type. In order to study the
Yuki Kitahara et al.
Bioscience, biotechnology, and biochemistry, 78(2), 336-342 (2014-07-19)
Rhodosporidium toruloides is a lipid-producing yeast, the growth of which is severely suppressed when hydrolysates of lignocellulosic biomass are used as carbon source. This is probably due to the toxic substances, such as organic acids, furans, and phenolic compounds produced
Giorgia Purcaro et al.
Analytical and bioanalytical chemistry, 407(1), 309-319 (2014-09-12)
The potentiality of a multidimensional comprehensive gas chromatographic (GC × GC) method, employing a simultaneous dual detection (FID and mass spectrometer), to generate peculiar two-dimensional chromatograms to be used as a chemical fingerprint, was investigated to characterize minor compounds in edible oil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service