Skip to Content
Merck
All Photos(2)

Key Documents

50490

Supelco

Methyl ethanesulfonate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Ethanesulfonic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SO3CH3
CAS Number:
Molecular Weight:
124.16
Beilstein:
1747452
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

CCS(=O)(=O)OC

InChI

1S/C3H8O3S/c1-3-7(4,5)6-2/h3H2,1-2H3

InChI key

YLJRCXSSKLWCDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl ethanesulfonate belongs to the group of alkyl sulfonate esters, termed as potential genotoxic impurities (GTIs).[1]
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Methyl ethanesulfonate may be used as certified reference material for the quantification of the analyte in pharmaceutical formulations using the gas chromatography technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

182.3 °F

Flash Point(C)

83.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a sensitive method for alkyl sulfonate genotoxic impurities determination in drug substances using gas chromatography coupled to triple quadrupole mass spectrometry
Liu Z, et al.
Journal of Pharmaceutical and Biomedical Analysis, 168(3), 23-29 (2019)
Yu Tang et al.
Talanta, 222, 121500-121500 (2020-11-11)
Alkylated DNA adducts are the most important and common form of DNA damage at the molecular level. In addition to known alkylated DNA adducts, many unknown DNA adducts remain to be discovered. A prediction-driven MRM profiling MS strategy has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service