Skip to Content
Merck
All Photos(1)

Documents

217468

Supelco

AmberChrom 50WX2 Ion Exchange Resin

hydrogen form, 100-200 mesh

Synonym(s):

AmberChrom 50WX2 hydrogen form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

product name

AmberChrom 50WX2 100-200 Mesh (H+) Cation Exchange Resin,

description

cross-linkage 2%

Quality Level

form

beads

parameter

150 °C max. temp.

moisture

74-82%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

operating pH

0-14

capacity

0.6 meq/mL by wetted bed volume

separation technique

cation exchange

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberchrom 50WX2 hydrogen form is a heat stable, water stable and low molecular weight cationic substance which is capable of purifying approximately 5000-fold by ion-exchange chromatography. Ion-exchange resins have good role as drug delivery systems.
Strongly acidic cation exchange resin. This ion-exchange resin has not been specially processed or cleaned. We suggest that it be subjected to a suitable preliminary elution and wash prior to use.

Application

Amberchrom 50WX2 hydrogen form was used to remove calcium from aqueous samples.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The purification and assay of the prostatic antibacterial factor (PAF).
W R Fair et al.
Biochemical medicine, 8(2), 329-339 (1973-10-01)
Seong Hoon Jeong et al.
International journal of pharmaceutics, 361(1-2), 26-32 (2008-06-21)
Complexes of ion-exchange resins and dextromethorphan (DM), a model drug, were prepared using different methods including a batch and a column method with different functional groups, ion-exchange capacity, degree of crosslinking, and resin particle size. Drug loading efficiency, release profiles
Sodium dodecyl sulphate-coated alumina for the extraction/preconcentration of benzimidazolic fungicides from natural waters prior to their quantification by liquid chromatography/fluorimetry.
Moral, Antonia, et al.
Analytica Chimica Acta, 569, 132-138 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service