Skip to Content
Merck
All Photos(1)

Key Documents

10103241001

Roche

Carnitine Acetyltransferase

from pigeon breast muscle

Synonym(s):

acetylcarnitine transferase

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204

biological source

pigeon breast

Quality Level

form

suspension

specific activity

~80 units/mg protein (At 25 °C with CoA and acetyl-D,L-carnitine as the substrates.)

packaging

pkg of 1 mL (5 mg)

manufacturer/tradename

Roche

technique(s)

analytical sample preparation: suitable

color

white

optimum pH

7.3-8.0

solubility

water: miscible

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

foreign activity

Acetyl-CoA-deacylase 0.01%

storage temp.

2-8°C

Gene Information

pigeon ... CRAT(102084317)

General description

Acetyl-CoA:carnitine O-acetyltransferase
Carnitine acetyltransferase is a mitochondrial enzyme essential for fuel utilization. It is present in the mitochondrial matrix and peroxisome. It is involved in the fatty acid metabolism. It facilitates conversion of acetyl-CoA and carnitine to acetylcarnitine and CoA respectively. This conversion facilitates peroxisomal β-oxidation by importing acetyl group for mitochondrial oxidation.

Quality

Contaminants: <0.01% acetyl-CoA-deacylase ion in storage buffer.

Physical form

Suspension in 2.9 M ammonium sulfate solution, 50 mM K-phosphate, 1 mM EDTA, pH approximately 7.5

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiankang Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(4), 1876-1881 (2002-02-21)
We test whether the dysfunction with age of carnitine acetyltransferase (CAT), a key mitochondrial enzyme for fuel utilization, is due to decreased binding affinity for substrate and whether this substrate, fed to old rats, restores CAT activity. The kinetics of
Y Elgersma et al.
The EMBO journal, 14(14), 3472-3479 (1995-07-17)
Carnitine acetyltransferase (CAT) is present in mitochondria and peroxisomes of oleate-grown Saccharomyces cerevisiae. Both proteins are encoded by the same gene, YCAT, which encodes a protein with a mitochondrial targeting signal (MTS) at the N-terminus, and a peroxisomal targeting signal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service