Skip to Content
Merck
All Photos(1)

Key Documents

SCM104

Sigma-Aldrich

3dGRO® R-Spondin-1 Conditioned Media Supplement

Alternative stem cell media supplement to purified recombinant R-Spondin-1 protein for organoid cultures.

Synonym(s):

3dGRO Media Supplement, Conditioned Media Supplement, R-Spondin-1 Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

General description

3D organoid culture systems are increasingly employed as powerful tools for the study of human diseases. R-Spondin proteins are a family of cysteine-rich, thrombospondin type I repeat containing proteins that activate both the WNT/β-catenin and WNT/PCP pathways. R-spondin-1 (RSPO1) is one of the most extensively used niche factors for culturing 3D organoids and has been used to establish organoids from the stomach, small intestine, colon, pancreas and liver from both mouse and human sources1,2,3 along with intestinal organoids from other large and small animals4. However, due to the high concentration requirement in organoid media the use of the recombinant R-spondin-1 becomes prohibitively expensive. The 3dGRO® R-Spondin-1 Conditioned Media Supplement is a potent and inexpensive alternative to purified recombinant RSPO1 protein for organoid cultures and is optimized for the highest level of RSPO-1 expression to ensure consistency and performance.



Source:


The 3dGRO® R-Spondin-1 Conditioned Media Supplement is generated from a 293T cell transfectant stably secreting mouse R-spondin-1 tagged with an influenza hemagglutinin (HA) epitope at the N-terminus and a murine IgG2a Fc fragment at the C-terminus. The conditioned medium does not contain serum.

Storage and Stability

RSPO-1 Conditioned Medium should be stored at -20°C. When ready to use, thaw the supplement overnight at 2-8°C in the dark. Thawed RSPO-1 Conditioned Medium is good for two weeks when stored at 2-8°C.

Legal Information

3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Organoid culture FAQs. Learn how to culture and analyze organoids following established 3D cell culture protocols. Tips and tricks.

Organoid culture FAQs. Learn how to culture and analyze organoids following established 3D cell culture protocols. Tips and tricks.

See All

Related Content

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Monitor barrier formation using colon PDOs, iPSC-derived colon organoids, Millicell® cell culture inserts, and the Millicell® ERS. 3.0.

Monitor barrier formation using colon PDOs, iPSC-derived colon organoids, Millicell® cell culture inserts, and the Millicell® ERS. 3.0.

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service