Skip to Content
Merck
All Photos(2)

Key Documents

S2HVU01RE

Millipore

Millipore® Stericup® Quick Release Vacuum Filtration System

pore size 0.45 μm, PVDF membrane, funnel capacity 150 mL, pack of 12 ea

Synonym(s):

SCHVU01RE, Stericup Quick Release-HV Sterile Vacuum Filtration System, sterile vacuum filter unit, sterile vacuum filtering unit, vacuum filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24
Pricing and availability is not currently available.

material

PVDF membrane
polystyrene bottle
polystyrene funnel(s)
polystyrene housing

Quality Level

sterility

sterile; irradiated

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

parameter

45 °C max. temp.

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

120 mm

capacity

150 mL

diam.

12.5 cm

filter diam.

73 mm

filtration area

40 cm2

funnel capacity

150 mL

matrix

Durapore®

pore size

0.45 μm pore size

fitting

inlet funnel
outlet flask (Receiver)

shipped in

ambient

General description

This item is a direct replacement for SCHVU01RE
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

Stericup® and Steritop® vacuum sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.

This Stericup® 0.45 μm pore size polyvinylidene fluoride (PVDF) membrane filter unit combines a 150 mL bottle-top with a 150 mL receiver flask and cap for processing and storage.

Application

Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–10 of 15 Questions  
  1. Can Stericup® bottles be placed inside freezers for long-term storage of media?

    1 answer
    1. You can successfully freeze and store many aqueous solutions (such as culture media) in Stericup® bottles at temperatures up to -20 °C (-4 °F). It is strongly recommended that you run a sample stability trial under your actual storage conditions prior to using Stericup® bottles for frozen storage.

      Helpful?

  2. Are Stericup® and Steritop® filters GMP-certified?

    1 answer
    1. No, Stericup® and Steritop® devices are not GMP-approved.

      Helpful?

  3. Can Stericup® and Steritop® filters be reused?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single use, disposable processing.

      Helpful?

  4. What leachables are detected during filtration processes using Stericup® filters?

    1 answer
    1. We have not performed extractables/leachables studies for our Stericup® filters.

      Helpful?

  5. What is the chemical compatibility of the various Stericup® and Steritop® filters?

    What is the chemical compatibility of the various Stericup® and Steritop® filters?

    1 answer
    1. Helpful?

  6. What is the difference between classic Stericup® filters and Stericup® “Quick Release” filters?

    1 answer
    1. Stericup® Quick Release, or “QR”, filters feature a connection between funnel filter and receiver bottle that requires only a quarter-turn release to facilitate funnel removal. These filters come fully engaged – there is no need to tighten before use. The receiver bottle cap has flanges on the top surface to improve grip, as well as a positive lock stop to ensure a leak-proof seal with the receiver bottle. Other improvements include a large, frosted writing surface on the receiver bottle and a lighter cap color to facilitate labelling, more prominent volume markings on the filtration funnel and receiver bottle, and a product label with a peel-off lot record strip on the bag for easy viewing in storage.

      Helpful?

  7. What is the expiration date of Stericup® and Steritop® filters?

    1 answer
    1. Typically 3 years from sterilization.

      Helpful?

  8. What Bacterial Endotoxins Test is used?

    1 answer
    1. The test method used complies with USP 85 and EP 2.6.14 to ensure the absence of endotoxin in our products.

      Helpful?

  9. What tubing can be used with Stericup® and Steritop® vacuum filters?

    What tubing can be used with Stericup® and Steritop® vacuum filters?

    1 answer
    1. Stericup® and Steritop® filters are fitted with a 1/4” tube adapter. We recommend 3/16 in. (4.8 mm) ID silicone tubing to fit this tube adapter: https://www.sigmaaldrich.com/product/mm/xx7100004

      Helpful?

  10. Can Stericup® and Steritop® filters be used to prepare solutions that will be administered to patients?

    1 answer
    1. No, all Stericup® and Steritop® devices are Research Use Only.

      Helpful?

1–10 of 15 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service