Skip to Content
Merck
All Photos(2)

Key Documents

MABN736

Sigma-Aldrich

Anti-NEGR1, clone 4H2.1 Antibody

clone 4H2.1, from mouse

Synonym(s):

Neuronal growth regulator 1, IgLON family member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
4H2.1, monoclonal
application:
IHC
WB
species reactivity:
rat, human
technique(s):
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4H2.1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NEGR1(257194)

General description

Neuronal growth regulator 1 (NEGR1) is also called IgLON family member 4. NEGR1 belongs to the immunoglobulin superfamily and is thought to be involved in cell-adhesion. NEGR1 possibly functions as a trans-neural growth-promoting factor in regenerative axon sprouting in the mammalian brain.

Immunogen

GST-tagged recombinant protein corresponding to human NEGR1.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected NEGR1 in human cerebellum and rat cerebral cortex tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-NEGR1, clone 4H2.1 Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of NEGR1.

Quality

Evaluated by Western Blotting in human hippocampus tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected NEGR1 in 10 µg of human hippocampus tissue lysate.

Target description

~50 kDa observed

Linkage

Replaces: MABN1072

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service