Skip to Content
Merck
All Photos(2)

Key Documents

MABD116

Sigma-Aldrich

Anti-Dux4 Antibody, clone 9A12

clone 9A12, from mouse

Synonym(s):

double homeobox protein 4, Double homeobox protein 10, Double homeobox protein 4/10, Dux-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9A12, monoclonal

species reactivity

human

technique(s)

electrophoretic mobility shift assay: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Dux4 is a homeodomain protein with a similar protein sequence to Pax3 and Pax7. Defects in Dux4 may be the cause of facioscapulohumeral muscular dystrophy (FSHD). FSHD is characterized by weakness of the muscles of the face, upper-arm and shoulder girdle. Ectopic expression of Dux4 in skeletal muscle activates the expression of stem cell and germline genes, and, when overexpressed in somatic cells, Dux4 can ultimately lead to cell death.

Immunogen

Epitope: domain of: C-Terminal region
recombinant protein corresponding to the domain of: C-Terminal region of Human Dux4.

Application

Detect Dux4 using this mouse monoclonal antibody, Anti-Dux4 Antibody, clone 9A12 validated for use in western blotting, ICC, Electrophoretic Mobility Shift Assay & IP.
Research Category
Stem Cell Research

Stem Cell Research
Research Sub Category
Muscle Physiology

Developmental Signaling
This antibody has been shown to work in immunocytochemistry applications (Dixit et al. 2007). This antibody has been shown to work in electrophoretic mobility shift assays (Dixit et al. 2007). This antibody has been shown to work in immunoprecipitation applications (Anseau et al., 2009).

Quality

Evaluated by Western Blot in a human skeletal muscle tissue lysate. Western Blot Analysis: A 1:250 dilution of this antibody detected Dux4 in a human skeletal muscle tissue lysate.

Target description

~58 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human skeletal muscle tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thuy-Hang Nguyen et al.
International journal of molecular sciences, 25(6) (2024-03-28)
FacioScapuloHumeral muscular Dystrophy (FSHD) is one of the most prevalent inherited muscle disorders and is linked to the inappropriate expression of the DUX4 transcription factor in skeletal muscles. The deregulated molecular network causing FSHD muscle dysfunction and pathology is not
Eugénie Ansseau et al.
Genes, 8(3) (2017-03-10)
FacioScapuloHumeral muscular Dystrophy (FSHD) is one of the most prevalent hereditary myopathies and is generally characterized by progressive muscle atrophy affecting the face, scapular fixators; upper arms and distal lower legs. The FSHD locus maps to a macrosatellite D4Z4 repeat
Louise A Moyle et al.
eLife, 5 (2016-11-15)
Facioscapulohumeral muscular dystrophy (FSHD) involves sporadic expression of DUX4, which inhibits myogenesis and is pro-apoptotic. To identify target genes, we over-expressed DUX4 in myoblasts and found that the receptor tyrosine kinase Ret was significantly up-regulated, suggesting a role in FSHD.
Céline Vanderplanck et al.
Skeletal muscle, 8(1), 2-2 (2018-01-14)
Facioscapulohumeral muscular dystrophy (FSHD) is associated with DNA hypomethylation at the 4q35 D4Z4 repeat array. Both the causal gene DUX4 and its homolog DUX4c are induced. DUX4c is immunodetected in every myonucleus of proliferative cells, while DUX4 is present in
Luyuan Li et al.
STAR protocols, 4(1), 102109-102109 (2023-03-01)
Transduction with lentiviral vectors is a useful approach to study the molecular function of specific genes in mammalian cells. Here, we present a calcium phosphate-based transfection protocol that guarantees highly efficient production and delivery of lentiviral vectors in adherent cultured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service