HCWG047S6
S-Pak® Membrane Filter
mixed cellulose esters (MCE), pore size 0.7 μm, filter diam. 47 mm, white gridded membranes for microbiological analysis of wastewater
Synonym(s):
Membrane filters, Semi-permeable membranes, S-Pak Filters 0.7µm 47mm white gridded
About This Item
Recommended Products
material
gridded filter
mixed cellulose esters (MCE)
white filter
Agency
ISO 7704
sterility
sterile; ethylene oxide treated
feature
hydrophilic
manufacturer/tradename
S-Pak®
diam.
47 mm
filter diam.
47 mm
pore size
0.7 μm pore size
input
sample type beverage(s)
sample type food(s)
sample type wine
sample type beer
sample type bottled water
sample type cosmetics
sample(s) (environmental)
sample type cider
sample type soft drink
sample type sport drink
application(s)
bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc
compatibility
for use with EZ-Fit® Filtration Manifold
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
S-Pak® membrane filters for microbial bioburden testing
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service