Skip to Content
Merck
All Photos(1)

Key Documents

HAWG047500

Millipore

MF-Millipore Membrane Filter, 0.45 µm pore size, gridded

MF-Millipore, filter diam. 47 mm, hydrophilic, white, gridded, pkg of 500 discs

Synonym(s):

MCE Membrane Filter, 0.45 μm Pore Size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.22

material

gridded
mixed cellulose esters (MCE) membrane
white filter

Quality Level

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, gridded, 500 discs

sterility

non-sterile

feature

hydrophilic

packaging

pkg of 500 discs

manufacturer/tradename

MF-Millipore
Millipore

filter diam.

47 mm

color

white

matrix

MF-Millipore

pore size

0.45 μm pore size

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods

Application

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, gridded, 500 discs

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service