Skip to Content
Merck
All Photos(1)

Key Documents

GPWP01300

Millipore

PES Membrane Filter, 0.22 μm Pore Size

greener alternative

Millipore Express®, filter diam. 13 mm, hydrophilic

Synonym(s):

Millipore Express PLUS Membrane Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

plain filter
polyethersulfone membrane
white filter

Quality Level

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Millipore Express®
Millipore

sustainability

Greener Alternative Product

parameter

15 mL/min water flow rate at 690 mbar (10 psi)

diam.

13 mm

filter diam.

13 mm

thickness

160-185 μm

impurities

≤0.5 EU/mL (LAL test, Aqueous extraction)

pore size

0.22 μm pore size

bubble point

≥1.38 bar, air with 100% IPA at 23 °C

greener alternative category

shipped in

ambient

General description

Filter Code: GPWP
Filter Type: Screen filter
The Millipore Express PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.

Features & Benefit:
•Ultra fast filtration and high throughput shorten process time
•Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications

Applications:
Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Sterile filtration
  • Buffer filtration
  • Tissue culture media filtration
  • Sample filtration for PFAS analytical testing (LC-MS)

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MILLIPORE EXPRESS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Validation of Millex® PES syringe filters in PFAS testing of water samples using EPA method 537.1.

Validation of Millex® PES syringe filters in PFAS testing of water samples using EPA method 537.1.

Validation of Millex® PES syringe filters in PFAS testing of water samples using EPA method 537.1.

Validation of Millex® PES syringe filters in PFAS testing of water samples using EPA method 537.1.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service