Skip to Content
Merck
All Photos(3)

Key Documents

EZBNZST-185K

Millipore

Benzonase® ST ELISA with ZooMAb Antibodies

Synonym(s):

Benzonase® Salt Tolerant endonuclease ELISA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158

parameter

50 μL

Quality Level

assay range

recovery range: 15.6 pg/mL
standard curve range: 15.6-1000 pg/mL

technique(s)

ELISA: suitable

size

96 well

input

cell culture supernatant

detection method

colorimetric

shipped in

wet ice

storage temp.

2-8°C

General description

ELISA for the specific detection of Benzonase ST (Salt Tolerant Benzonase).

Application

96 stripwell formatt ELISA

Features and Benefits

Specifically detects Benzonase® Salt Tolerant Nuclease using a pair of specifc recombinant monoclonal ZooMAb® Rabbit antibodies in a sandwich ELISA formatt. Results are determined spectrophotometrically using a Benzonase® ST standard curve. The assay range is 15.6-1000pg ml, with an LLOQ of 15.6pg (%CV/%Recovery ≤20%).

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service