Skip to Content
Merck
All Photos(1)

Key Documents

ABE70

Sigma-Aldrich

Anti-BAF180 Antibody

from rabbit

Synonym(s):

polybromo 1, protein polybromo-1, BRG1-associated factor 180, polybromo-1D, Polybromo-1D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ChIP
WB
species reactivity:
human, mouse, rat
technique(s):
ChIP: suitable
western blot: suitable
citations:
8

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PBRM1(55193)

General description

BRG1-associated factor 180 (BAF180) is a subunit of the SWI/SNF-B (PBAF) chromatin remodeling complex, and is known to contain 6 bromo domains, 2 BAH domains, and a single HMG box DNA-binding domain. BAF180 has a critical and active role in the transcriptional activation and repression of specific genes via chromatin remodeling, including genes involved in the development of the heart, thus suggesting a role in mammalian development. Recent studies demonstrate BAF180 to suppress tumorigenesis through its ability to regulate both baseline and signal-dependent p21 transcription by its binding of the p21 promoter. Expression of BAF180 is reported in a wide range of tissues.

Immunogen

GST-tagged recombinant protein corresponding to human BAF180.

Application

Anti-BAF180 Antibody is a Rabbit Polyclonal Antibody for detection of BAF180 also known as protein polybromo-1, BRG1-associated factor 180 & has been validated in WB.
Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in ChIP (Xia, W., et al. (2008).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected BAF180 in 10 µg of HEK293 cell lysate.

Target description

~193 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darmood Wei et al.
Genes, chromosomes & cancer, 59(8), 472-483 (2020-04-08)
Renal medullary carcinoma (RMC) is a rare, aggressive disease that predominantly afflicts individuals of African or Mediterranean descent with sickle cell trait. RMC comprises 1% of all renal cell carcinoma diagnoses with a median overall survival of 13 months. Patients are
Alena Shmakova et al.
Cells, 10(6) (2021-07-03)
PBRM1, a component of the chromatin remodeller SWI/SNF, is often deleted or mutated in human cancers, most prominently in renal cancers. Core components of the SWI/SNF complex have been shown to be important for the cellular response to hypoxia. Here
Christopher J Petell et al.
The Journal of biological chemistry, 299(8), 104996-104996 (2023-07-03)
A critical component of gene regulation is recognition of histones and their post-translational modifications by transcription-associated proteins or complexes. Although many histone-binding reader modules have been characterized, the bromo-adjacent homology (BAH) domain family of readers is still poorly characterized. A
Lili Liao et al.
Molecular oncology, 13(4), 811-828 (2018-12-27)
Polybromo-1 (PBRM1) is an important tumor suppressor in kidney cancer. It contains six tandem bromodomains (BDs), which are specialized structures that recognize acetyl-lysine residues. While BD2 has been found to bind acetylated histone H3 lysine 14 (H3K14ac), it is not
Ryan J Conrad et al.
Molecular cell, 67(6), 1001-1012 (2017-08-29)
BET proteins commonly activate cellular gene expression, yet inhibiting their recruitment paradoxically reactivates latent HIV-1 transcription. Here we identify the short isoform of BET family member BRD4 (BRD4S) as a corepressor of HIV-1 transcription. We found that BRD4S was enriched in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service