Skip to Content
Merck
All Photos(2)

Key Documents

ABE135

Sigma-Aldrich

Anti-Sp1 Antibody

from rabbit

Synonym(s):

Sp1 transcription factor, specificity protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
IP
WB
species reactivity:
mouse, human, rat
technique(s):
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
7
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SP1(6667)

General description

Sp1 was one of the first transcription factors to be characterized in mammalian cells. The Sp1 protein plays a role in the regulation of gene expression. This protein has three Cys2His2 zinc-finger motifs that have been shown to bind to the consensus sequence 5′-(G/T)GGGCGG(G/A)(G/A)(C/T)-3′ also known as a GC box. Sp1 is a ubiquitous transcription factor required for the expression of a variety of genes, such as those involved in the cell cycle and development. Interaction of the Sp1 protein with the GC box activates transcription of specific mRNAs from genes that contain functional recognition sites.

Specificity

Homology to chimpanzee, canine, and rhesus monkey is 99%.
This antibody recognizes Sp1.

Immunogen

GST-tagged recombinant protein corresponding to full-length human Sp1.

Application

Anti-Sp1 Antibody is a Rabbit Polyclonal Antibody for detection of Sp1 also known as Sp1 transcription factor, specificity protein 1 & has been validated in WB, IP, ICC.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Sp1 in HeLa and A431 cells.

Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated Sp1 from 500 µg of HeLa nuclear extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 0.1 µg/mL of this antibody detected Sp1 on 10 µg of HeLa nuclear extract.

Target description

95 kDa and 105 kDa

Linkage

Replaces: MABE550

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrien Morel et al.
Oncotarget, 8(28), 46163-46176 (2017-05-19)
High-risk human papillomaviruses are the etiological agents of cervical cancer and HPV16 is the most oncogenic genotype. Immortalization and transformation of infected cells requires the overexpression of the two viral oncoproteins E6 and E7 following HPV DNA integration into the
K A Gajewska et al.
Nature communications, 12(1), 5904-5904 (2021-10-10)
The importin superfamily member Importin-13 is a bidirectional nuclear transporter. To delineate its functional roles, we performed transcriptomic analysis on wild-type and Importin-13-knockout mouse embryonic stem cells, revealing enrichment of differentially expressed genes involved in stress responses and apoptosis regulation.
Lina Adwan et al.
Neuropharmacology, 79, 596-602 (2014-01-28)
Environmental exposure to lead (Pb) early in life results in a latent upregulation of genes and products associated with Alzheimer's disease (AD), particularly the plaque forming protein amyloid beta (Aβ). Furthermore, animals exposed to Pb as infants develop cognitive decline
Christoph Hoffmann et al.
PloS one, 8(12), e83426-e83426 (2014-01-07)
Uncoupling protein (UCP) 3 is a mitochondrial inner membrane protein implicated in lipid handling and metabolism of reactive oxygen species. Its transcription is mainly regulated by peroxisome proliferator-activated receptors (PPAR), a family of nuclear hormone receptors. Employing bandshift assays, RNA
Sandra Kissing et al.
Autophagy, 13(4), 670-685 (2017-01-28)
The vacuolar-type H+-translocating ATPase (v-H+-ATPase) has been implicated in the amino acid-dependent activation of the mechanistic target of rapamycin complex 1 (MTORC1), an important regulator of macroautophagy. To reveal the mechanistic links between the v-H+-ATPase and MTORC1, we destablilized v-H+-ATPase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service