Skip to Content
Merck
All Photos(1)

Key Documents

AB15504

Sigma-Aldrich

Anti-Klotho Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... KL(9365)

General description

Animal models of human aging, which display the characteristic diseases associated with aging, provide insight into the cause of such diseases. Recently a transgenic mouse model, called Klotho, with several premature aging phenotypes has been described. They display premature aging phenotypes such as Osteoporosis, age related skin changes, ectopic calcifications, atrophy of genital organs and thymus, emphysema and short life span (Kuro-o et al., 1997). KL protein associated with the Klotho mutation is a 1014 amino acid long peptide. It has a putative signal sequence at its N-terminus and a single transmembrane domain near its C-terminus, which is postulated to anchor it to the membrane (Martin & Mian, 1997). The extracellular domain has two internal repeats, KL-1 and KL-2 which have 20-40% sequence identity to bacterial b-glucosidase of plants and animals and mammalian lactase glycosylceramidase suggesting a role for Klotho in sphingolipid metabolism (Martin & Mian, 1997). kl gene expression was observed to be tissue specific. Improvement of systemic aging phenotypes in kl/kl mice occurs even when the exogenous expression was limited to some organs, suggesting that KL associated aging is regulated through a humoral signaling pathway. KL has been reported to be localized on the cell surface when expressed on Cho cells (Kuro-o et al., 1997). Human kl cDNA is expected to encode a protein of 1012 amino acids and has 86% homology to mouse kl.

Specificity

Klotho
Other species have not yet been tested. The immunogen sequence is 82% conserved in human and 76% in rat klotho.

Immunogen

Synthetic peptide near the C-terminus of mouse Klotho.

Application

Detect Klotho using this Anti-Klotho Antibody validated for use in ELISA & WB.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Yan et al.
Molecular medicine reports, 22(2), 1035-1043 (2020-05-30)
Klotho is a type of single‑pass transmembrane protein that is important for the proper function of numerous organs. The aim of the present study was to investigate the role of Klotho in sepsis‑associated myocardial damage. In the present study, reverse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service