Skip to Content
Merck
All Photos(1)

Key Documents

8.03181

Sigma-Aldrich

Benzyl acetate

for synthesis

Synonym(s):

Benzyl acetate, Acetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
205-399-7
NACRES:
NA.22
form:
liquid
Assay:
≥99.0% (GC)

vapor pressure

1 hPa ( 45 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

460 °C

potency

2490 mg/kg LD50, oral (Rat)

expl. lim.

0.9-8.4 % (v/v)

bp

205-207 °C/1013 hPa

mp

-51 °C

transition temp

flash point 95 °C

density

1.06 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(Cc1ccccc1)C(=O)C

InChI

1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QUKGYYKBILRGFE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.055 - 1.056
Identity (IR): passes test

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service