8.01667
1-Bromopropane
for synthesis
Synonym(s):
1-Bromopropane, Propyl bromide
About This Item
Recommended Products
vapor pressure
146 hPa ( 20 °C)
Quality Level
Assay
≥99.0% (GC)
form
liquid
autoignition temp.
490 °C
potency
>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)
expl. lim.
3.4-9.1 % (v/v)
bp
71 °C/1013 hPa
mp
-108 °C
transition temp
flash point -10 °C
solubility
2.5 g/L
density
1.35 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
BrCCC
InChI
1S/C3H7Br/c1-2-3-4/h2-3H2,1H3
InChI key
CYNYIHKIEHGYOZ-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.352 - 1.355
Identity (IR): passes test
Signal Word
Danger
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Ozone 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
71.6 °F - closed cup
Flash Point(C)
22 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service