Skip to Content
Merck
All Photos(1)

Key Documents

8.00918

Sigma-Aldrich

Dimethyl phthalate

for synthesis

Synonym(s):

Dimethyl phthalate, Phthalic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(COOCH3)2
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
12352108
EC Index Number:
205-011-6
NACRES:
NA.22
grade:
for synthesis
bp:
283 °C/1013 hPa
vapor pressure:
0.008 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor pressure

0.008 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

460 °C (DIN 51794)

potency

5120 mg/kg LD50, oral (Rat)
>12000 mg/kg LD50, skin (Rabbit)

dilution

(for synthesis)

pH

7 (20 °C in H2O)

bp

283 °C/1013 hPa

mp

6 °C

transition temp

flash point 150 °C

solubility

4.0 g/L

density

1.19 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)c1c(cccc1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3

InChI key

NIQCNGHVCWTJSM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.189 - 1.192
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service