Skip to Content
Merck
All Photos(1)

Key Documents

5.31551

Sigma-Aldrich

PKR/PERK Activator, DHBDC

Synonym(s):

PKR/PERK Activator, DHBDC, Protein Kinase R (PKR) Activator, Protein Kinase R Like Kinase (PERK) Activator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O6
CAS Number:
Molecular Weight:
312.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

~10 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to pink

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell permeable chromenone derivative that acts as a dual activator of protein kinase R (PKR) and protein kinase R like kinase (PERK). Induces the phosphorylation of IkBa and thereby activates NF-κB pathway. However, the activation of NF-κB is independent of PKR activity. Induces elF2a phosphorylation and activates its downstream effectors CHOP/GADD153. Does not affect the phosphorylation of Akt, MAP kinase, or 4E-BP1. Inhibits the expression of cyclin D1 and E and blocks the growth of CRL-2813 and MCF-7 cells (IC50 = ~ 10 µM). Also shown to reduce JFH1 hepatitis C virus infectivity (~40% inhibition at 5 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKR and PERK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bai, H., et al. 2013. ChemBioChem14, 1255.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service