Skip to Content
Merck
All Photos(1)

Key Documents

5.09703

Sigma-Aldrich

LSD1 Inhibitor VII, SP-2509

Synonym(s):

LSD1 Inhibitor VII, SP-2509, BHC110 Inhibitor VII, Histone Lysine Demethylase Inhibitor XIV, KIAA1718 Inhibitor VII, LSD Inhibitor VII, SP-2509

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20ClN3O5S
CAS Number:
Molecular Weight:
437.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C19H20ClN3O5S/c1-13(17-12-15(20)5-6-18(17)24)21-22-19(25)14-3-2-4-16(11-14)29(26,27)23-7-9-28-10-8-23/h2-6,11-12,24H,7-10H2,1H3,(H,22,25)/b21-13+

InChI key

NKUDGJUBIVEDTF-FYJGNVAPSA-N

General description

A cell-permeable phenethylidene-benzohydrazide compound that acts as a potent, selective, reversible, and non-competitive inhibitor of Lysine-Specific Demethylase 1 (LSD1; IC50 = 13 nM; Ki = 31 nM). Exhibits excellent selectivity over monoamine oxidases A and B (IC50 >300 µM), lactate dehydrogenase, glucose oxidase, hERG, CYP1A2, CYP2D6 (IC50 ≥ 10 µM), CYP2C9 (IC50 = 8.04 µM) and CYP2C19 (IC50 = 9.76 µM). Inhibits CYP3A4 only at >200-fold higher concentration. Reduces the proliferation of several cancer cell lines, including AN3 Ca, BT-20, MCF-7, T-47D, HT29, MIA PaCa-2 and SK-N-MC (IC50 = 356, 489, 637, 649, 429, 468 and 329 nM, respectively).
A cell-permeable, lysine-specific demethylase 1 (LSD1) active site-targeting phenethylidene-benzohydrazide that inhibits LSD1 activity (IC50 = 13 nM) in a reversible and substrate non-competitive (Ki = 34 nM) manner, while inhibiting CYP3A4 only at much higher concentrations (IC50 = 2.61 µM) and displaying little or no potency towards CYP1A2/2C9/2C19/2D6. MAO-A/B, D-lactate dehydrogenase, glucose oxidase, and hERG (IC50 ≥8.0 µM). Shown to enhance histone H3 Lys9 dimethylation (H3K9me2) in androgen-dependent prostate cancer VCaP cultures (1 to 10 µM) and effectively inhibits LSD1-dependent cancer growth (IC50 in nM = 329/SK-N-MC, 356/AN3 Ca, 429/HT29, 468/MIA PaCa-2, 489/BT-20, 612/HER218, 614/HCT 116, 637/MCF-7, 649/T-47D; 96 h).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LSD1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Fiskus, W., et al. 2014. Leukemia28, 2155.
Sorna, V., et al. 2013. J. Med. Chem.56, 9496.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service