Skip to Content
Merck
All Photos(1)

Key Documents

219261

Sigma-Aldrich

Catalase, Aspergillus niger

Catalase, Aspergillus niger, CAS 9001-05-2, is a native, long-acting, extremely stable form of catalase that is active over a wide pH range (pH 1 - 12).

Synonym(s):

Catalase, Aspergillus niger

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 1900

form

lyophilized powder

specific activity

≥10,000 units/mg protein
≥4000 units/mg dry wt.

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

green

solubility

H2O: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O(CC)C(=O)c1ccc(cc1)O

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

General description

Research area: Cell Signaling

Catalase, Aspergillus niger, is a native, long-acting, extremely stable form of catalase that is active over a wide pH range (pH 1 - 12). Composed of four protein subunits, each containing a heme group bound to its active site, it is one of the most important antioxidant enzymes present in almost all aerobic organisms.[1]

Application

Catalase may be used in:
  • Industrial applications for eliminating hydrogen peroxide, which is utilized as an oxidizing, bleaching, or sterilizing agent.
  • Food industry and egg processing, often in conjunction with other enzymes.
  • Food preservation methods in combination with glucose oxidase, where superoxide dismutase produces H2O2, which is subsequently eliminated by catalase.
  • Preservation process, along with glucose oxidase.

Biochem/physiol Actions

Catalase is responsible for breaking down two hydrogen peroxide molecules into one oxygen molecule and two water molecules in a two-step reaction. It plays a crucial role in regulating the cellular level and catabolism of hydrogen peroxide which in turn shields the cells from oxidative damage. Its malfunctioning and deficiency is linked to various diseases, including vitiligo, diabetes mellitus, anemia, cardiovascular diseases, bipolar disorders, Wilson disease, certain dermatological disorders, Alzheimer′s disease, hypertension, and schizophrenia. Anomalies in catalase activity are related to acatalasemia or Takahara disease, a rare genetic disorder.[1]

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will decompose 1.0 µmol H₂O₂ per min at 30°C, pH 7.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Note: 1 KU = 1000 units

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of Catalase in Oxidative Stress- and Age-Associated Degenerative Diseases
Nandi A, et al.
Oxidative Medicine and Cellular Longevity, 9613090-9613090 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service