Skip to Content
Merck
All Photos(1)

Key Documents

1.46006

Millipore

FREY Agar

suitable for microbiology, For avian mycoplasma detection, pkg of 60 mm plates (20 plates per box)

Synonym(s):

Mycoplasma Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.75

Agency

EP 2.6.7
USP

Quality Level

sterility

sterile; aseptically filled

form

plate

feature

ready-to-use

packaging

pkg of 60 mm plates (20 plates per box)

pH

7.8 ( in H2O)

application(s)

industrial qc
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

mycoplasma

General description

FREY Agar is supplemented with swine serum to provide assimilable cholesterol and fatty acids, and NAD allows the growth of M. synoviae. Phenol red in the medium changes the color of the medium to yellow or purple, indicating the growth of mycoplasmas. Furthermore, penicillin inhibits the growth of accompanying bacterial flora.

Application

FREY Agar is recommended for the detection of avian mycoplasmas in pharmaceutical products, especially in bulk vaccines, cell banks, and virus cultures.

Features and Benefits

  • Ready-to-use convenience as no further preparation steps are required.
  • Pharmaceutical products such as Biopharmaceuticals, Vaccines, Cell cultures, and Virus cultures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service