Skip to Content
Merck
All Photos(2)

Key Documents

1.06647

Supelco

Sodium sulfate

anhydrous 99.99 Suprapur®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-820-9
NACRES:
NB.21
Assay:
99.99%
grade:
for inorganic trace analysis
form:
solid
solubility:
200 g/L
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

Assay

99.99%

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

solubility

200 g/L

density

2.68 g/mL at 25 °C (lit.)

bulk density

1400‑1600 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
719591.06649793531
solubility

200 g/L

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

-

solubility

-

assay

99.99%

assay

≥99.0% (T)

assay

≥99.0% (alkalimetric), 98.5-101% dry basis (alkalimetric)

assay

≥99%

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

100

grade

for inorganic trace analysis

grade

anhydrous

grade

ACS reagent

grade

anhydrous

mp

884 °C (lit.)

mp

884 °C (lit.)

mp

884 °C (lit.)

mp

884 °C (lit.)

Application

  • Iron Phosphate Purification: Sodium sulfate is involved in methods for removing impurities from chemical compounds. A recent study focused on developing a technique to remove sodium sulfate from iron phosphate, which is crucial for producing high-purity materials for various industrial applications. This research highlights sodium sulfate′s role in refining chemical processes and improving product quality (Li et al., Advances in Analytical Chemistry, 2022).

Analysis Note

Purity (metallic): ≥ 99.99 %
Assay (acidimetric): ≥ 99.5 %
Chloride (Cl): ≤ 5 ppm
Total nitrogen (N): ≤ 5 ppm
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 5.0 ppm
Bi (Bismuth): ≤ 0.005 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.005 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.010 ppm
Cs (Cesium): ≤ 5 ppm
Cu (Copper): ≤ 0.005 ppm
Eu (Europium): ≤ 0.005 ppm
Fe (Iron): ≤ 0.050 ppm
K (Potassium): ≤ 5 ppm
La (Lanthanum): ≤ 0.005 ppm
Li (Lithium): ≤ 0.1 ppm
Mg (Magnesium): ≤ 0.05 ppm
Mn (Manganese): ≤ 0.010 ppm
Ni (Nickel): ≤ 0.005 ppm
Pb (Lead): ≤ 0.005 ppm
Rb (Rubidium): ≤ 1.0 ppm
Sc (Scandium): ≤ 0.005 ppm
Sm (Samarium): ≤ 0.005 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.005 ppm
Y (Yttrium): ≤ 0.005 ppm
Yb (Ytterbium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.020 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service