Skip to Content
Merck
All Photos(1)

Key Documents

1.04169

Sigma-Aldrich

Glycine

buffer substance for electrophoresis

Synonym(s):

Gly, Aminoacetic acid, Aminoethanoic acid, Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
MDL number:
UNSPSC Code:
41105305
EC Index Number:
200-272-2
NACRES:
NC.01
technique(s):
electrophoresis: suitable
vapor pressure:
0.0000171 Pa ( 25 °C)
Pricing and availability is not currently available.

vapor pressure

0.0000171 Pa ( 25 °C)

Assay

≥99.7% (perchloric acid titration)

form

solid

potency

7930 mg/kg LD50, oral (Rat)

technique(s)

electrophoresis: suitable

pH

5.9-6.4 (20 °C, 50 g/L in H2O)

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

solubility

soluble 250 g/L

density

1.161 g/cm3 at 20 °C

bulk density

920 kg/m3

storage temp.

15-25°C

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Analysis Note

Assay (perchloric acid titration): ≥ 99.7 %
Identity (IR-spectrum): passes test
Chloride (Cl): ≤ 0.003 %
Heavy metals (as Pb): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.02 %
Foreign amino acids: ≤ 0.1 %
Other ninhydrine positive substances: ≤ 0.1 %
Suitability (as buffer substance for electrophoresis): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service