Skip to Content
Merck
All Photos(4)

Key Documents

06-172-I

Sigma-Aldrich

Anti-Na+/K+ ATPase alpha-3

from rabbit

Synonym(s):

ATP1A3, Na+/K+ ATPase alpha-3, Sodium/potassium-transporting ATPase subunit alpha-3, Na(+)/K(+) ATPase alpha-3 subunit, Na(+)/K(+) ATPase alpha(III) subunit, Sodium pump subunit alpha-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Sodium/potassium-transporting ATPase subunit alpha-3 (UniProt: P06687; also known as EC: 3.6.3.9, Na(+)/K(+) ATPase alpha-3 subunit, Na(+)/K(+) ATPase alpha(III) subunit, Sodium pump subunit alpha-3) is encoded by the Atp1a3 gene (Gene ID: 24213) in rat. Sodium/potassium-transporting ATPase subunit alpha-3 is a multi-pass membrane protein that is exclusive to the brain tissue and is highly expressed in cell bodies and projections of hippocampus. It is responsible for establishing and maintaining the electrochemical gradients of sodium and potassium ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. Sodium/potassium-transporting ATPase subunit alpha-3 is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The catalytic component catalyzes the hydrolysis of ATP coupled with the exchange of sodium and potassium ions across the plasma membrane. Reduced levels of neuronal sodium/ potassium ATPase activity in ATP1A3 heterozygous mice increases their susceptibility to depression-like endophenotypes by chronic variable stress.

Specificity

This rabbit polyclonal antibody detects Sodium/potassium-transporting ATPase subunit alpha-3 in human, mouse, and rat brain. It targets an epitope with in 25 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal region of rat Sodium/potassium-transporting ATPase subunit alpha-3.

Application

Anti-Na+/K+ ATPase alpha-3, Cat. No. 06-172-I, is a highly specific rabbit polyclonal antibody that targets Sodium/potassium-transporting ATPase subunit alpha-3 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250-1,000 dilution of this antibody detected Na+/K+ ATPase alpha-3 in human cerebral cortex and human cardiac muscle tissue.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in adult human brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Na+/K+ ATPase alpha-3 in 10 µg of adult human brain tissue lysate.

Target description

~112 kDa observed; 111.75 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bavat Bornstein et al.
eLife, 12 (2023-02-07)
The proprioceptive system is essential for the control of coordinated movement, posture, and skeletal integrity. The sense of proprioception is produced in the brain using peripheral sensory input from receptors such as the muscle spindle, which detects changes in the
Komal Sodhi et al.
iScience, 24(11), 103262-103262 (2021-11-11)
Recent studies suggest that a western diet may contribute to clinical neurodegeneration and dementia. Adipocyte-specific expression of the Na,K-ATPase signaling antagonist, NaKtide, ameliorates the pathophysiological consequences of murine experimental obesity and renal failure. In this study, we found that a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service